siRNA Details
| Gene Symbol: Luc7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025881.3 | NP_080157.1 | 3 | 337 | ||
| NM_028190.3 | NP_082466.2 | 3 | 337 | |||
| GenBank mRNA | AK007781.1 | - | 4 | 361 | ||
| AK010539.1 | - | 3 | 289 | |||
| AK017655.1 | - | 3 | 297 | |||
| AK160293.1 | - | 3 | 338 | |||
| AK211801.1 | - | 4 | 265 | |||
| BC019194.1 | - | 4 | 330 | |||
| BC034135.1 | - | 2 | 503 | 5003664 | ||
| BC055875.1 | - | 3 | 1320 | |||
| BY726224.1 | - | 4 | 395 | |||

Luc7l
Genomic Map