siRNA Details
| Gene Symbol: Zcchc18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001035509.1 | NP_001030586.1 | 4 | 912 | ||
| NM_001035510.1 | NP_001030587.1 | 2 | 1383 | |||
| NM_025893.2 | NP_080169.2 | 4 | 860 | |||
| GenBank mRNA | AK005317.1 | - | 4 | 838 | ||
| AK075607.1 | - | 1 | 464 | |||
| AK134856.1 | - | 1241 | ||||
| AY650116.1 | - | 1 | 342 | |||
| BC017627.1 | - | 4 | 773 | 4505033 | ||

Zcchc18
Genomic Map