siRNA Details
| Gene Symbol: Psmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080554.2 | NP_542121.2 | 4 | 493 | ||
| GenBank mRNA | AK005335.1 | - | 4 | 471 | ||
| AK011026.1 | - | 4 | 470 | |||
| AK029067.1 | - | 3 | 790 | |||
| AK032992.1 | - | 4 | 471 | |||
| AK078419.1 | - | 4 | 471 | |||
| AK145587.1 | - | 4 | 470 | |||
| AK148284.1 | - | 494 | ||||
| AK220321.1 | - | 4 | 434 | |||
| BC019112.1 | - | 4 | 504 | |||

Psmd5
Genomic Map