siRNA Details
| Gene Symbol: Ttc23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168475.1 | NP_001161947.1 | 8 | 1436 | ||
| NM_001168476.1 | NP_001161948.1 | 7 | 1236 | |||
| NM_025905.3 | NP_080181.2 | 9 | 1562 | |||
| GenBank mRNA | AK005431.1 | - | 7 | 1197 | ||
| AK086049.1 | - | 1504 | ||||
| AK141754.1 | - | 8 | 1422 | |||
| BC038179.1 | - | 9 | 1533 | 4485073 | ||

Ttc23
Genomic Map