siRNA Details
| Gene Symbol: Mettl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025907.3 | NP_080183.1 | 5 | 873 | ||
| GenBank mRNA | AK005448.1 | - | 5 | 928 | ||
| AK006715.1 | - | 4 | 780 | |||
| AK078185.1 | - | 5 | 927 | |||
| AK082949.1 | - | 5 | 929 | |||
| AK133735.1 | - | 5 | 905 | |||
| AK189650.1 | - | 1 | 133 | |||
| BC030449.1 | - | 5 | 874 | 5400506 | ||

Mettl6
Genomic Map