siRNA Details
| Gene Symbol: Ccdc91 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025911.2 | NP_080187.2 | 10, 11 | 1131 | ||
| GenBank mRNA | AK007917.1 | - | 10, 11 | 1132 | ||
| BC006734.1 | - | 10, 11 | 1036 | 3156328 | ||
| BC024946.1 | - | 5, 6 | 389 | 4010938 | ||
| BC049073.1 | - | 10, 11 | 1100 | 5707404 | ||

Ccdc91
Genomic Map