siRNA Details
| Gene Symbol: Tbc1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194334.2 | NP_919315.2 | 2, 3 | 569 | ||
| GenBank mRNA | AK089175.1 | - | 2, 3 | 534 | ||
| AK129273.1 | - | 2, 3 | 555 | |||
| AK148382.1 | - | 2, 3 | 572 | |||
| AK156788.1 | - | 2, 3 | 556 | |||
| BY741512.1 | - | 2, 3 | 569 | |||

Tbc1d2b
Genomic Map