siRNA Details
Gene Symbol: Fam210b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016210.1 | - | 2 | 386 | 4924329 | |
BC057642.1 | - | 2 | 373 | 2655111 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam210b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC016210.1 | - | 2 | 386 | 4924329 | |
BC057642.1 | - | 2 | 373 | 2655111 |