siRNA Details
| Gene Symbol: Tmem88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025915.4 | NP_080191.3 | 1 | 103 | ||
| GenBank mRNA | AK010789.1 | - | 1 | 109 | ||
| AK011240.1 | - | 1 | 104 | |||
| AK045149.1 | - | 1 | 104 | |||
| AK161789.1 | - | 1 | 104 | |||
| BC100316.1 | - | 1 | 92 | |||
| BC120912.2 | - | 1 | 104 | 40091214 | ||

Tmem88
Genomic Map