siRNA Details
| Gene Symbol: Thap4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025920.3 | NP_080196.3 | 2 | 942 | ||
| GenBank mRNA | AK086483.1 | - | 2 | 756 | ||
| AK140888.1 | - | 885 | ||||
| AK167814.1 | - | 2 | 702 | |||
| BC057963.1 | - | 1 | 240 | |||
| BC063758.1 | - | 2 | 673 | 6334411 | ||
| BC066042.1 | - | 2 | 968 | 6413779 | ||

Thap4
Genomic Map