siRNA Details
| Gene Symbol: Rbm25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027349.3 | NP_081625.3 | 10 | 1335 | ||
| GenBank mRNA | AK038317.1 | - | 1 | 2637 | ||
| AK142132.1 | - | 4 | 611 | |||
| AK162135.1 | - | 1432 | ||||
| BC158104.1 | - | 9 | 1215 | 9007388 | ||

Rbm25
Genomic Map