siRNA Details
| Gene Symbol: Rbm25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_027349.3 | NP_081625.3 | 7, 8 | 927 | ||
| GenBank mRNA | AK011403.1 | - | 7, 8 | 908 | ||
| AK142132.1 | - | 1, 2 | 203 | |||
| AK151910.1 | - | 7, 8 | 879 | |||
| AK162135.1 | - | 1020 | ||||
| AK164367.1 | - | 7, 8 | 892 | |||
| BC010792.1 | - | 6, 7 | 673 | |||
| BC023749.1 | - | 5, 6 | 636 | |||
| BC158104.1 | - | 6, 7 | 807 | 9007388 | ||

Rbm25
Genomic Map