siRNA Details
| Gene Symbol: Oxct1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024188.6 | NP_077150.1 | 10 | 1341 | ||
| GenBank mRNA | AB085609.1 | - | 10 | 1083 | ||
| AK009027.1 | - | 10 | 1121 | |||
| AK011354.1 | - | 10 | 1021 | |||
| AK078086.1 | - | 10 | 1339 | |||
| AK146158.1 | - | 10 | 1099 | |||
| AK146342.1 | - | 10 | 1086 | |||
| AK151662.1 | - | 10 | 1080 | |||
| AK151695.1 | - | 10 | 1071 | |||
| AK167681.1 | - | 10 | 1171 | |||
| AK215681.1 | - | 3 | 197 | |||
| BC003422.1 | - | 10 | 1045 | 2598824 | ||

Oxct1
Genomic Map