siRNA Details
| Gene Symbol: Pus3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023292.4 | NP_075781.3 | 3 | 493 | ||
| GenBank mRNA | AF266505.1 | - | 2 | 447 | ||
| AK030639.1 | - | 3 | 889 | |||
| AK079095.1 | - | 3 | 494 | |||
| BC029253.1 | - | 2 | 454 | 4187198 | ||

Pus3
Genomic Map