siRNA Details
| Gene Symbol: Slc25a53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001082412.2 | NP_001075881.1 | 7 | 779 | ||
| NM_001114179.1 | NP_001107651.1 | 2 | 161 | |||
| GenBank mRNA | AK076279.1 | - | 7 | 774 | ||
| CX240158.1 | - | 2 | 161 | |||

Slc25a53
Genomic Map