siRNA Details
| Gene Symbol: Romo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164216.1 | NP_001157688.1 | 3 | 422 | ||
| NM_001164217.2 | NP_001157689.1 | 3 | 436 | |||
| NM_025946.6 | NP_080222.1 | 2 | 520 | |||
| GenBank mRNA | AA058111.1 | - | 3 | 422 | ||
| AK003488.1 | - | 3 | 331 | |||
| AK008300.1 | - | 3 | 357 | |||
| AY028425.1 | - | 2 | 415 | |||
| BC028749.1 | - | 3 | 317 | 1196408 | ||
| CJ221444.1 | - | 3 | 384 | |||

Romo1
Genomic Map