siRNA Details
| Gene Symbol: Pi4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025951.3 | NP_080227.2 | 8 | 1268 | ||
| NM_028744.3 | NP_083020.2 | 8 | 1199 | |||
| GenBank mRNA | AF411321.1 | - | 8 | 1123 | ||
| AK011751.1 | - | 8 | 1231 | |||
| AK016754.1 | - | 8 | 1162 | |||
| AK035523.1 | - | 8 | 1253 | |||
| AK170730.1 | - | 8 | 1252 | |||
| AY148879.1 | - | 1162 | ||||
| BC010257.1 | - | 276 | ||||
| BC062144.1 | - | 8 | 1175 | 6335661 | ||

Pi4k2b
Genomic Map