siRNA Details
| Gene Symbol: Pi4k2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025951.3 | NP_080227.2 | 4 | 838 | ||
| NM_028744.3 | NP_083020.2 | 4 | 769 | |||
| GenBank mRNA | AF411321.1 | - | 4 | 693 | ||
| AK011751.1 | - | 4 | 801 | |||
| AK035523.1 | - | 4 | 823 | |||
| AK170730.1 | - | 4 | 822 | |||
| BC062144.1 | - | 4 | 745 | 6335661 | ||

Pi4k2b
Genomic Map