siRNA Details
| Gene Symbol: Magt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190409.1 | NP_001177338.1 | 1, 2 | 244 | ||
| NM_025952.4 | NP_080228.4 | 1, 2 | 244 | |||
| GenBank mRNA | AK010320.1 | - | 1, 2 | 180 | ||
| AK012185.1 | - | 1, 2 | 121 | |||
| AK013243.1 | - | 1, 2 | 125 | |||
| AK018623.1 | - | 1, 2 | 147 | |||
| AK136622.1 | - | 1, 2 | 126 | |||
| AK144696.1 | - | 1, 2 | 123 | |||
| BC003881.1 | - | 1, 2 | 131 | 3496572 | ||
| DQ000005.1 | - | 1, 2 | 244 | |||

Magt1
Genomic Map