siRNA Details
| Gene Symbol: Tex40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039494.2 | NP_001034583.1 | 5 | 609 | ||
| GenBank mRNA | AK005644.1 | - | 5 | 588 | ||
| AK006034.1 | - | 5 | 547 | |||
| AK006134.1 | - | 5 | 612 | |||
| AK007089.1 | - | 5 | 585 | |||
| BC048452.1 | - | 5 | 546 | |||
| BC061075.1 | - | 5 | 551 | |||
| BC172038.1 | - | 4 | 797 | |||
| BC172041.1 | - | 4 | 852 | |||
| BC172042.1 | - | 4 | 766 | |||

Tex40
Genomic Map