siRNA Details
Gene Symbol: 1700019D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144953.2 | NP_659202.1 | 2 | 784 | ||
GenBank mRNA | AK010796.1 | - | 2 | 785 | ||
AK010944.1 | - | 2 | 646 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700019D03Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144953.2 | NP_659202.1 | 2 | 784 | ||
GenBank mRNA | AK010796.1 | - | 2 | 785 | ||
AK010944.1 | - | 2 | 646 |