siRNA Details
| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025961.5 | NP_080237.1 | 4 | 649 | ||
| GenBank mRNA | AK007325.1 | - | 4 | 649 | ||
| AK146052.1 | - | 4 | 648 | |||
| AK146111.1 | - | 4 | 650 | |||
| AK151313.1 | - | 4 | 648 | |||
| AK152069.1 | - | 4 | 650 | |||
| AK153450.1 | - | 4 | 634 | |||
| AY625267.1 | - | 2 | 232 | |||

Gatm
Genomic Map