siRNA Details
| Gene Symbol: Gatm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025961.5 | NP_080237.1 | 2 | 276 | ||
| GenBank mRNA | AK007325.1 | - | 2 | 276 | ||
| AK145862.1 | - | 2 | 277 | |||
| AK145914.1 | - | 2 | 277 | |||
| AK146052.1 | - | 2 | 275 | |||
| AK146111.1 | - | 2 | 277 | |||
| AK151313.1 | - | 2 | 275 | |||
| AK152069.1 | - | 2 | 277 | |||
| AK153450.1 | - | 2 | 262 | |||
| BC003879.1 | - | 2 | 281 | 3495871 | ||

Gatm
Genomic Map