siRNA Details
| Gene Symbol: Mmachc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025962.3 | NP_080238.2 | 4 | 795 | ||
| GenBank mRNA | AK007725.1 | - | 4 | 730 | ||
| AK012761.1 | - | 4 | 732 | |||
| AK084194.1 | - | 4 | 746 | |||
| BC054756.1 | - | 4 | 734 | 6411797 | ||

Mmachc
Genomic Map