siRNA Details
| Gene Symbol: 2310039H08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025966.3 | NP_080242.1 | 1 | 747 | ||
| GenBank mRNA | AK003321.1 | - | 1 | 560 | ||
| AK009696.1 | - | 1 | 570 | |||
| BC038674.1 | - | 1 | 747 | 1513865 | ||
| BU055991.1 | - | 1 | 542 | |||

2310039H08Rik
Genomic Map