siRNA Details
| Gene Symbol: Zbtb8os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025970.3 | NP_080246.1 | 7 | 474 | ||
| GenBank mRNA | AK007377.1 | - | 7 | 464 | ||
| AK008007.1 | - | 7 | 475 | |||
| AK132279.1 | - | 485 | ||||
| AY071852.1 | - | 7 | 463 | |||
| BC026442.1 | - | 469 | ||||
| BC037657.1 | - | 444 | ||||
| BC046825.1 | - | 481 | ||||
| BC094629.1 | - | 7 | 505 | 3376726 | ||
| BC125606.1 | - | 7 | 475 | 40130121 | ||
| BC125608.1 | - | 7 | 475 | 40130123 | ||

Zbtb8os
Genomic Map