siRNA Details
| Gene Symbol: Zfp787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013012.1 | NP_001013030.1 | 3 | 837 | ||
| GenBank mRNA | AK077458.1 | - | 3 | 804 | ||
| AK159396.1 | - | 2 | 662 | |||
| BC137753.1 | - | 3 | 759 | 8860772 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp787| Gene Symbol: Zfp787 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013012.1 | NP_001013030.1 | 3 | 837 | ||
| GenBank mRNA | AK077458.1 | - | 3 | 804 | ||
| AK159396.1 | - | 2 | 662 | |||
| BC137753.1 | - | 3 | 759 | 8860772 | ||