siRNA Details
| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177552.1 | NP_001171023.1 | 5 | 917 | ||
| NM_025976.5 | NP_080252.1 | 7 | 1292 | |||
| GenBank mRNA | AK013874.1 | - | 7 | 1157 | ||
| AK028592.1 | - | 5 | 756 | |||
| AK041839.1 | - | 6 | 1031 | |||
| BC027221.1 | - | 7 | 1133 | 4196466 | ||

Bfar
Genomic Map