siRNA Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290454.1 | NP_001277383.1 | 4 | 446 | ||
| NM_023305.3 | NP_075794.3 | 4 | 446 | |||
| GenBank mRNA | AF275549.3 | - | 4 | 405 | ||
| AK009596.1 | - | 4 | 405 | |||
| AK028477.1 | - | 4 | 405 | |||
| AK028670.1 | - | 4 | 397 | |||
| AK030370.1 | - | 4 | 387 | |||
| AK033121.1 | - | 4 | 398 | |||
| AK076078.1 | - | 4 | 405 | |||
| AK163229.1 | - | 4 | 404 | |||
| BC065996.1 | - | 4 | 394 | 5689710 | ||

Ubap1
Genomic Map