siRNA Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025982.4 | NP_080258.1 | 1, 2 | 140 | ||
| GenBank mRNA | AK012457.1 | - | 1, 2 | 141 | ||
| AK012567.1 | - | 1, 2 | 143 | |||
| AK012853.1 | - | 1, 2 | 143 | |||
| AK013523.1 | - | 1, 2 | 153 | |||
| AK014183.1 | - | 1, 2 | 139 | |||
| AK075962.1 | - | 1, 2 | 141 | |||
| AK160269.1 | - | 1, 2 | 97 | |||
| BC023306.1 | - | 1, 2 | 117 | 4165774 | ||

Tspan31
Genomic Map