siRNA Details
| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025985.4 | NP_080261.2 | 5 | 780 | ||
| GenBank mRNA | AK012539.1 | - | 5 | 687 | ||
| AK013902.1 | - | 5 | 661 | |||
| AK083029.1 | - | 5 | 723 | |||
| AK167472.1 | - | 5 | 646 | |||
| AK209284.1 | - | 2 | 71 | |||
| BC065083.1 | - | 6 | 874 | |||
| BC096474.1 | - | 897 | 4503077 | |||

Ube2g1
Genomic Map