siRNA Details
| Gene Symbol: Ube2g1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025985.4 | NP_080261.2 | 4 | 724 | ||
| GenBank mRNA | AK012539.1 | - | 4 | 631 | ||
| AK013902.1 | - | 4 | 605 | |||
| AK083029.1 | - | 4 | 667 | |||
| AK167472.1 | - | 4 | 590 | |||
| AK209284.1 | - | 1 | 14 | |||
| BC065083.1 | - | 5 | 818 | |||
| BC096474.1 | - | 841 | 4503077 | |||
| DV657669.1 | - | 4 | 724 | |||

Ube2g1
Genomic Map