siRNA Details
| Gene Symbol: MAGEA12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001166386.3 | NP_001159858.1 | 3 | 665 | ||
| NM_001166387.3 | NP_001159859.1 | 3 | 573 | |||
| NM_005367.6 | NP_005358.2 | 2 | 467 | |||
| GenBank mRNA | AK314168.1 | - | 3 | 665 | ||
| BC003408.2 | - | 2 | 455 | 3450217 | ||
| BC098146.1 | - | 3 | 245 | |||
| BQ436888.1 | - | 2 | 467 | |||
| BT007108.1 | - | 1 | 311 | |||
| CR541775.1 | - | 1 | 311 | |||

MAGEA12
Genomic Map