siRNA Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 21 | 2688 | ||
| GenBank mRNA | AK147960.1 | - | 21 | 2611 | ||
| AK149648.1 | - | 21 | 2680 | |||
| AK161293.1 | - | 9 | 1294 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herc6| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 21 | 2688 | ||
| GenBank mRNA | AK147960.1 | - | 21 | 2611 | ||
| AK149648.1 | - | 21 | 2680 | |||
| AK161293.1 | - | 9 | 1294 | |||