siRNA Details
| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 16, 17 | 2108 | ||
| GenBank mRNA | AK147960.1 | - | 16, 17 | 2031 | ||
| AK149648.1 | - | 16, 17 | 2100 | |||
| AK161293.1 | - | 4, 5 | 714 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Herc6| Gene Symbol: Herc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025992.2 | NP_080268.1 | 16, 17 | 2108 | ||
| GenBank mRNA | AK147960.1 | - | 16, 17 | 2031 | ||
| AK149648.1 | - | 16, 17 | 2100 | |||
| AK161293.1 | - | 4, 5 | 714 | |||