siRNA Details
| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025995.2 | NP_080271.2 | 2 | 610 | ||
| GenBank mRNA | AK011820.1 | - | 2 | 608 | ||
| AK160201.1 | - | 2 | 566 | |||
| BC053434.1 | - | 2 | 534 | 6336178 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fbxo5| Gene Symbol: Fbxo5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025995.2 | NP_080271.2 | 2 | 610 | ||
| GenBank mRNA | AK011820.1 | - | 2 | 608 | ||
| AK160201.1 | - | 2 | 566 | |||
| BC053434.1 | - | 2 | 534 | 6336178 | ||