siRNA Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289524.1 | NP_001276453.1 | 5 | 597 | ||
| NM_001289525.1 | NP_001276454.1 | 3 | 344 | |||
| NM_024194.6 | NP_077156.2 | 5 | 600 | |||
| GenBank mRNA | AK011726.1 | - | 5 | 581 | ||
| AK017814.1 | - | 5 | 569 | |||
| AK034978.1 | - | 580 | ||||
| AK148748.1 | - | 5 | 559 | |||
| AK152244.1 | - | 3 | 325 | |||
| AK163288.1 | - | 5 | 568 | |||
| AK170052.1 | - | 5 | 577 | |||

Lrrc40
Genomic Map