siRNA Details
| Gene Symbol: Nkain1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025998.3 | NP_080274.2 | 2, 3 | 339 | ||
| GenBank mRNA | AK011164.1 | - | 2, 3 | 472 | ||
| AK011850.1 | - | 2, 3 | 270 | |||
| BC116391.1 | - | 2, 3 | 269 | 40101865 | ||
| BC116392.1 | - | 2, 3 | 269 | |||
| CJ161499.1 | - | 2, 3 | 339 | |||
| EF058048.2 | - | 2, 3 | 392 | |||

Nkain1
Genomic Map