siRNA Details
| Gene Symbol: Sft2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026006.1 | NP_080282.1 | 1 | 1987 | ||
| GenBank mRNA | AK011898.1 | - | 1 | 1362 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sft2d3| Gene Symbol: Sft2d3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026006.1 | NP_080282.1 | 1 | 1987 | ||
| GenBank mRNA | AK011898.1 | - | 1 | 1362 | ||