siRNA Details
| Gene Symbol: Cdt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026014.3 | NP_080290.3 | 6 | 991 | ||
| GenBank mRNA | AB086655.1 | - | 6 | 970 | ||
| AF477481.1 | - | 6 | 991 | |||
| AK012041.1 | - | 6 | 968 | |||
| AK017609.1 | - | 6 | 965 | |||
| AK028287.1 | - | 6 | 963 | |||
| AK049163.1 | - | 6 | 963 | |||
| AK076527.1 | - | 5 | 1073 | |||
| AK083015.1 | - | 6 | 966 | |||
| BC013842.1 | - | 6 | 946 | |||
| BC024485.1 | - | 3 | 242 | |||
| BC048076.1 | - | 6 | 991 | 6307476 | ||

Cdt1
Genomic Map