siRNA Details
| Gene Symbol: Zmat5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026015.2 | NP_080291.1 | 6 | 655 | ||
| GenBank mRNA | AK007565.1 | - | 6 | 642 | ||
| AK012098.1 | - | 6 | 656 | |||
| AK013415.1 | - | 5 | 628 | |||
| BC031796.1 | - | 6 | 634 | 4205598 | ||

Zmat5
Genomic Map