siRNA Details
| Gene Symbol: Zmynd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026021.3 | NP_080297.1 | 5 | 684 | ||
| GenBank mRNA | AJ409150.1 | - | 5 | 645 | ||
| AJ409151.1 | - | 5 | 530 | |||
| AK005908.1 | - | 5 | 672 | |||
| AK012484.1 | - | 5 | 685 | |||
| AK088063.1 | - | 5 | 705 | |||
| AK184476.1 | - | 1 | 48 | |||
| BC027321.1 | - | 5 | 650 | 3990966 | ||

Zmynd19
Genomic Map