siRNA Details
| Gene Symbol: Zcrb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026025.2 | NP_080301.1 | 5 | 419 | ||
| GenBank mRNA | AB095989.1 | - | 5 | 452 | ||
| AK012589.1 | - | 6 | 502 | |||
| AK012852.1 | - | 5 | 420 | |||
| AK170136.1 | - | 5 | 431 | |||
| BC058368.1 | - | 5 | 453 | 6819507 | ||

Zcrb1
Genomic Map