siRNA Details
| Gene Symbol: Rnpc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038696.1 | NP_001033785.1 | 5 | 600 | ||
| NM_001287015.1 | NP_001273944.1 | 5 | 600 | |||
| NM_026043.3 | NP_080319.2 | 5 | 600 | |||
| GenBank mRNA | AK013288.1 | - | 5 | 544 | ||
| AK134241.1 | - | 5 | 565 | |||
| AK161063.1 | - | 5 | 583 | |||
| AK161974.1 | - | 5 | 553 | |||
| AK163729.1 | - | 5 | 553 | |||
| AK173272.1 | - | 5 | 351 | |||
| AK187884.1 | - | 5 | 403 | |||
| BC016603.1 | - | 2 | 116 | 3587176 | ||
| BC043695.1 | - | 5 | 569 | 5250774 | ||

Rnpc3
Genomic Map