siRNA Details
| Gene Symbol: Tmem19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133683.3 | NP_598444.1 | 2 | 282 | ||
| GenBank mRNA | AK018383.1 | - | 2 | 231 | ||
| AK034298.1 | - | 2 | 288 | |||
| AK036228.1 | - | 1 | 359 | |||
| AK042426.1 | - | 1 | 337 | |||
| AK136778.1 | - | 271 | ||||
| AK138931.1 | - | 2 | 270 | |||
| AK155275.1 | - | 2 | 243 | |||
| AK170143.1 | - | 2 | 242 | |||
| AK171087.1 | - | 2 | 242 | |||
| AK209903.1 | - | 1 | 28 | |||
| AK213890.1 | - | 1 | 28 | |||
| BC016895.1 | - | 2 | 284 | 4239007 | ||

Tmem19
Genomic Map