siRNA Details
| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026045.3 | NP_080321.2 | 6 | 643 | ||
| GenBank mRNA | AK035073.1 | - | 6 | 673 | ||
| AK135707.1 | - | 6 | 683 | |||
| AK135890.1 | - | 6 | 664 | |||
| AK139720.1 | - | 6 | 631 | |||
| AK161550.1 | - | 2 | 87 | |||
| BC004573.1 | - | 6 | 643 | 3586127 | ||
| BC028306.1 | - | 6 | 646 | |||
| BC058631.1 | - | 6 | 638 | 6402639 | ||

Prpf18
Genomic Map