siRNA Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023324.2 | NP_075813.2 | 5 | 743 | ||
| GenBank mRNA | AF302503.1 | - | 4 | 965 | ||
| AK151660.1 | - | 5 | 744 | |||
| AK152900.1 | - | 5 | 744 | |||
| AK152997.1 | - | 5 | 743 | |||
| AK153372.1 | - | 5 | 744 | |||
| AK167525.1 | - | 5 | 744 | |||
| BC016515.1 | - | 4 | 342 | 3600130 | ||
| BC026695.1 | - | 5 | 707 | |||

Peli1
Genomic Map