siRNA Details
| Gene Symbol: 2810474O19Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289661.1 | NP_001276590.1 | 4 | 4360 | ||
| NM_001289662.1 | NP_001276591.1 | 3 | 4273 | |||
| NM_026054.3 | NP_080330.1 | 3 | 4291 | |||
| GenBank mRNA | AK220403.1 | - | 3 | 4274 | ||
| BC038922.1 | - | 1 | 500 | |||
| BC055761.1 | - | 1 | 368 | |||
| BC086679.1 | - | 1 | 3711 | |||
| BC089381.1 | - | 1 | 311 | |||
| BC139435.1 | - | 3 | 4213 | 8862457 | ||
| BC139436.1 | - | 4 | 4300 | 8862458 | ||

2810474O19Rik
Genomic Map