siRNA Details
Gene Symbol: Zfp422 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF281634.1 | - | 2 | 390 | ||
AK082970.1 | - | 2 | 293 | |||
AK152681.1 | - | 2 | 277 | |||
AK169251.1 | - | 2 | 539 | |||
BC018339.1 | - | 1 | 262 | 3583940 | ||
BY082381.1 | - | 2 | 315 |
