siRNA Details
| Gene Symbol: Zfp422 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF281634.1 | - | 2 | 419 | ||
| AK082970.1 | - | 2 | 322 | |||
| AK152681.1 | - | 2 | 306 | |||
| AK169251.1 | - | 2 | 568 | |||
| AK178961.1 | - | 1 | 31 | |||
| AK187023.1 | - | 1 | 32 | |||
| AK212218.1 | - | 1 | 31 | |||
| AK214724.1 | - | 1 | 31 | |||
| BC018339.1 | - | 1 | 291 | 3583940 | ||
| BY082381.1 | - | 2 | 344 | |||

Zfp422
Genomic Map